Order Entry
Northern Ireland
ContactUsLinkComponent
(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (by HPLC), white to off-white solid
Irritant
(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (by HPLC), white to off-white solid
Catalog # ENZOBMLEA1040005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (by HPLC), white to off-white solid
Catalog # ENZOBMLEA1040005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥99% (HPLC)
  • Identity:
    Determined by H-NMR

Specification Test Results

About this item

 :
 :197.127 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:79055-68-8
Formula:C₅H₁₂NO₅P
MDL Number:MFCD00078839