Order Entry
Northern Ireland
ContactUsLinkComponent
Geldanamycin ≥98% (by TLC) (conjugated to FITC)
Geldanamycin ≥98% (by TLC) (conjugated to FITC)
Catalog # ENZOBMLEI3610001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Geldanamycin ≥98% (by TLC) (conjugated to FITC)
Catalog # ENZOBMLEI3610001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :560.64 g/mol
Storage Temperature:Freezer
CAS Number:30562-34-6
Formula:C₂₉H₄₀N₂O₉
MDL Number:MFCD00274570