Order Entry
Northern Ireland
ContactUsLinkComponent
Melatonin ≥98% (by HPLC)
Melatonin ≥98% (by HPLC)
Catalog # ENZOBMLNS5200001
Supplier:  Enzo Life Sciences
CAS Number:  
Melatonin ≥98% (by HPLC)
Catalog # ENZOBMLNS5200001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.28 g/mol
Storage Temperature:Freezer
CAS Number:73-31-4
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00005655