About this item
Non-reactive antibody to use as a negative control for mouse IgG1 primary antibodies. Available with CF® Dyes and other labels.
- Isotype control for antibody-based experiments
- Available with bright and photostable CF® Dyes
- APC, R-PE, RPE-Astral™616, APC-Astral™813, and biotin conjugates also available
- Suitable for western, flow cytometry, ELISA, immunofluorescence, and immunohistology
This is a non-reactive mouse monoclonal IgG1 kappa antibody labeled with one of our bright and photostable CF® Dyes and other labels. This antibody may serve as a control for a mouse monoclonal IgG1 antibody under identical experimental conditions. Biotium also carries mouse monoclonal isotype control antibodies for IgG2a Kappa (MG2a-53) and IgG2b Kappa (IGG1/1333).
Type: Isotype control
Antigen:
Clonality: Monoclonal
Clone:
Conjugation: CF®405L
Epitope:
Host: Mouse
Isotype: IgG1, Kappa
Reactivity:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- BNC511331-250
- BNC511331-1ML
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405L
- CF® 405L
- Host:
- Mouse
- Mouse
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 1 - 2 ug/ml
- 1 - 2 ug/ml
- Storage temperature:
- Store at 2...8 °C
- Store at 2...8 °C