- Filtration area:3,9 cm²
- Pore size:0,22 µm
- Sterility:Sterile
- Volume:100 ml
- Max. temperature:45 °C
- Colour:Clear/blue
- Description:GS syringe filter, 25 mm, MCE membrane, vented PVC housing, EO sterilized
- Hold-up volume:<0,1 ml
- Max. operating pressure:5,2 bar (75 psi)
- Pk:50
- Inlet/outlet connections:Female Luer-Lok/Male Luer slip
Vented Millex®-GS filter units are syringe filters for sterilisation of low volume aqueous solutions where venting is desired.
- Clarification of water, buffers or aqueous solutions
- Mixed Cellulose Esters (MCE)
This single-use product consists of a 0,22 μm hydrophilic MF-MilliporeTM mixed cellulose esters (MCE) membrane filter sealed in a PVC housing with a female Luer-Lok™ inlet connector and a male Luer-slip outlet connector. A 0,03 μm hydrophobic PTFE filter in the dome prevents air locking by automatically venting air introduced upstream.
Media: MF-Millipore
Sterility: Sterile
Sterilisation: Ethylene oxide
Wettability: Hydrophilic
Device material: PVC
Stem cell type: Mouse embryonic stem cells, Human embryonic stem cells, Mesenchymal stem cells, Neural stem cells, Hematopoietic stem cells, Epithelial cells, Pancreatic stem cells, Cardiac stem cells, Induced pluripotent stem cells.