Order Entry
Northern Ireland
ContactUsLinkComponent
Gauzes with Stainless Steel Centre
Gauzes with Stainless Steel Centre
Catalog # USBE5072
Gauzes with Stainless Steel Centre
Catalog # USBE5072

Specifications

  • Description:
    Gauze with stainless steel centre, 155×155 mm
  • Pk:
    1

Specifications

About this item

Mesh from galvanised steel wire.

  • Alternative to gauze with ceramic centre
  • Compatible with Usbeck 4-feet stand

Fireproof stainless steel centre made of 1,4841 in format 60×60 mm, prevents burner flame penetration.

Weight: 40 g