Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen name:Phytochrome
- Antigen symbol:Phy
- Clonality:Monoclonal
- Clone:Oat-8
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Pisum sativum,Avena sativa
- Western blot:Yes
- Format:Liquid
- Purity:Immunoglobulin Protein G purified, in PBS pH 7,4, 150 mM NaCl, 0,05% of sodium azide
- Molecular weight:124 kDa
- Storage temperature:Store at −20 °C; Avoid repeated freeze-thaw cycles. Please remember to spin the tubes briefly prior to opening them to avoid any losses that might occur from material adhering to the cap or sides of the tube.
- Immunogen:Phytochrome
- Pk:100 µG
Phytochrome is a photomorphogenically active pigment that modulates plant growth and development with respect to incident light intensity and wavelength distribution. It exists in two forms: an inactive, red-absorbing form (Pr),4 and an active far-red-absorbing form (Pfr). When either absorbs light, it is photoconverted to the other.
- This antibody can also be used in competitive ELISA
- Epitope recognised by this antibody is located ca. 70 kDa from N-terminus
- Recommended dilution: Assay dependent
Phytochrome is a dimeric, water-soluble, relatively labile chromoprotein with similar, if not identical, monomers of about 124 kDa each. It is also a relatively low abundance protein, even under the best of conditions. Genetic manipulation of phytochrome expression in plants leads to plants requiring less light and able to divert more energy to the production of fruits and seeds.
For its physicochemical characterization, it has therefore been difficult to utilize techniques that require large quantitites of highly purified protein. Consequently, indirect methods for elucidating its structure/function relationships are especially important. These could also be applicable to fabaceae and closely related families.
Type:
Antigen: Phy
Clonality: Monoclonal
Clone: Oat-8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Avena sativa, Pisum sativum