Pesticide reference standard, Endrin 100 µg/ml in methanol
Catalog # ACCUP-045S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:72-20-8
- Description:Endrin 100 µg/ml in methanol
- Formula:C₁₂H₈Cl₆O
- Description:Endrin