

Cesium hydroxide hydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12260-45-6
:MFCD00010964
Formula:HCsO·H₂O
:167.93 g/mol
Merck Index:14,2013
Storage Temperature:Ambient
EINECS:244-344-1
UN:2682
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together