

3-Nitro-p-toluonitrile ≥98%
Catalog # L18269.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:939-79-7
:
Formula:C₈H₆N₂O₂
:162.15
Boiling Pt:171 °C (12 mmHg)
Melting Pt:103…108 °C
UN:3439
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00031482