Order Entry
Northern Ireland
ContactUsLinkComponent
L-Alanine ≥99%, white crystalline powder
L-Alanine ≥99%, white crystalline powder
Supplier:  MP Biomedicals
Certificates
L-Alanine ≥99%, white crystalline powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Optical Rotation:
    +13,5° to +15,5°
  • Heavy Metals:
    ≤20 ppm

Specification Test Results

Frequently Bought Together