L-Alanine
Catalog # MOLEM58544402
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
CAS Number:56-41-7
Formula:C₃H₇NO₂
MDL Number:MFCD00064410