Order Entry
Northern Ireland
ContactUsLinkComponent
Pedal Stand for MAUSER WIVA Vat-5 Medical Containers
Pedal Stand for MAUSER WIVA Vat-5 Medical Containers
Catalog # DANIUK80800130
Pedal Stand for MAUSER WIVA Vat-5 Medical Containers
Catalog # DANIUK80800130

Specifications

  • W×D×H:
    426×485×886 mm
  • Pk:
    1
  • Weight:
    7 kg

Specifications

About this item

This mobile holder is suitable for MAUSER WIVA Infinity and WIVA Vat-5 containers of 30, 50 and 60 L.

  • Made of stainless steel
  • Comes with four rollers, including two lockable

Height with open lid: 1120 mm