Avery Address Labels, UltraGrip
DIALAVL7162
- Colour:White
- Description:Address labels
- Dimensions:99,1×34 mm
- Pk:1.600
With these easy to use Ultragrip labels, you can address envelopes in no time at all.
- For use with laser printers
- Quick peel technology for easy use and increased productivity
- Made from FSC certified paper from responsibly managed forests
- Jam free printing
QuickPEEL system, simply divides the labels along the perforation lines to expose the edges, peel and stick to your envelope. Perfect for envelopes, they are fully compatible with your laser printer and guarantee a jam free print for instant, professional labelling. These eco-friendly labels can be fully recycled and are made using environmentally sound materials. This pack contains 250 sheets with 16 labels per sheet (4000 labels in total).