Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SARS-CoV-2 (COVID-19) Spike S2 Llama Recombinant Monoclonal Antibody [P1B8]
Anti-SARS-CoV-2 (COVID-19) Spike S2 Llama Recombinant Monoclonal Antibody [P1B8]
  PRSISD9787
Anti-SARS-CoV-2 (COVID-19) Spike S2 Llama Recombinant Monoclonal Antibody [P1B8]
  PRSISD9787

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SARS-CoV-2 (COVID-19) Spike S2
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Llama
  • Reactivity:
    Virus
  • Form:
    Liquid
  • Gene ID:
    HEK293
  • Storage buffer:
    SARS-CoV-2 (COVID-19) Spike S2 Antibody is supplied in PBS
  • Storage temperature:
    Store at −20 °C, stable for up to one year.
  • Concentration:
    1 mg/ml
  • Immunogen:
    SARS-CoV-2 Spike S2 protein containing C-terminal His Tag. The protein was expressed in human 293 cells (HEK293). It contains amino acids Ser 686 - Pro 1213
  • Purification:
    SARS-CoV-2 (COVID-19) Spike S2 Antibody is affinity chromatography purified via Nickel column. Antibody is supplied as a His-tagged purified protein. It also contains a myc-tag for detection.
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: SARS-CoV-2 (COVID-19) Spike S2
Clonality: Recombinant Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype: sdAb
Reactivity: Virus