To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Label printer, colour, BradyJet J4000, UK plug
- Model:J4000-UK
- Print resolution:4800 dpi
- Materials supported:Continuous and die-cut
- Computer requirements:PC
- Print width:203,20 mm (max.)
- Power supply:AC adaptor
- Connections:USB; ethernet
- Printer:Full Colour
- Print method:Inkjet
- Print speed:5000 max. labels/day
- Pk:1
- For:Facility identification; general identification; hazard communication identification; lean and 5S labelling; pipe and valve marking; warehouse marking
Specifications
About this item
High-impact, precision full colour that lasts. Create the compliant, long-lasting labels, signs and tags you need with the BradyJet J4000 inkjet printer.
- Full colour
- Long-lasting
- 5000 labels per day
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material. Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colour options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels.
Streamline storage by only needing to stock white label material and print only what you need, when you need it, in photo-quality full colour every time. Easily print PDFs or from third-party software packages with the optimised windows driver. Add Brady workstation safety and facility ID software suite (J4000-BWSSFID) or lab suite (J4000-BWSLAB) to your solution and make label creation even easier.