ContactUsLinkComponent
Sodium hexanitritocobaltate(III) ACS, Fluka™
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III) ACS, Fluka™
Certificates
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III) ACS, Fluka™
Specification Test Results
  • Appearance of the substance:
    complying
  • Insoluble in acetic acid:
    Max. 0.020 %
  • Suitability for K determination:
    complying