


Boron tribromide 1 M in dichloromethane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-33-4
:
Formula:BBr₃
MDL Number:MFCD00011312
:250.52 g/mol
Merck Index:14,1347
Density:1,467
EINECS:233-657-9
UN:3390
ADR:6.1,I
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together