Order Entry
Northern Ireland
ContactUsLinkComponent
Pyridine-2,4-dicarboxylic acid, Sigma-Aldrich®
Irritant
Pyridine-2,4-dicarboxylic acid, Sigma-Aldrich®
Catalog # 8.21052.0010
Supplier:  Merck
CAS Number:  
Irritant
Pyridine-2,4-dicarboxylic acid, Sigma-Aldrich®
Catalog # 8.21052.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    2.49 g/l (25 °C)
  • Melting Point:
    242 - 243 °C
  • Molar Mass:
    167.12 g/mol

Specification Test Results

About this item

 :
 :167.12 g/mol
Boiling Pt:357 °C (1013 hPa)
Melting Pt:246…248 °C
Density:0,942 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-892-2
 :GHS07
CAS Number:499-80-9
 :MFCD00006296
Formula:C₇H₅NO₄
MDL Number:MFCD00149413