
1-Ethyl-3-methylimidazolium dicyanamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:370865-89-7
:MFCD28145696
Formula:C₈H₁₁N₅
MDL Number:MFCD08276373
:177.21 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together