
4-Bromo-3,5-xylenol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7463-51-6
:
Formula:C₈H₉BrO
MDL Number:MFCD00002315
Boiling Pt:259 °C (1013 hPa)
Melting Pt:113…117 °C
:201.06 g/mol
Storage Temperature:Ambient
EINECS:231-255-8
:GHS07
Related Information
Frequently Bought Together