


N,N-Diethylethylenediamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-36-7
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008176
:116.21 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:–70 °C
Density:0,82 g/cm³ (20 °C)
Flash Pt:36 °C
Storage Temperature:Ambient
UN:2685
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
EINECS:202-844-7
:GHS02
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-70 °C
- Molar Mass:116.21 g/mol
- Boiling Point:145 - 148 °C (1013 hPa)
- Flash Point:36 °C
- Explosion Limit:0.7 - 8.4 %(V)
- Density:0.82 g/cm3 (20 °C)
- Ignition Point:245 °C
Specification Test Results
Frequently Bought Together