
2,6-Dimethoxy-p-cresol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6638-05-7
:
Formula:C₉H₁₂O₃
MDL Number:MFCD00017289
:168.19 g/mol
Boiling Pt:144…146 °C (12 mmHg)
Melting Pt:36…42 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:229-641-6
:GHS07
Related Information
Frequently Bought Together