Order Entry
Northern Ireland
ContactUsLinkComponent
2,6-Dibromo-p-cresol ≥98%
Irritant
2,6-Dibromo-p-cresol ≥98%
Catalog # A18634.14
CAS Number:  
Irritant
2,6-Dibromo-p-cresol ≥98%
Catalog # A18634.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

CAS Number:2432-14-6
 :
Formula:C₇H₆Br₂O
 :265.93 g/mol
Boiling Pt:268…271 °C (1013 hPa)
Melting Pt:49…50 °C
Density:2,22 g/cm³ (20 °C)
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:219-404-5
 :GHS07
MDL Number:MFCD00002154