Anti-PKC theta Rabbit Polyclonal Antibody
NOVUNBP2-55717
- Antigen symbol:PKC theta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Antigen synonyms:PRKCT|nPKC-theta|MGC141919|MGC126514|PRKCQ|protein kinase C theta type|EC 2.7.11|EC 2.7.11.13|protein kinase C, theta
- Purity:Affinity purified
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at − 20 °C long term. Avoid freeze-thaw cycles.
- Size:100 µl
- Pk:100 µl
Type:
Antigen: PKC theta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat