ContactUsLinkComponent
Hydrometer for Lime Milk
Hydrometer for Lime Milk
  HECH46199010
New Product
Hydrometer for Lime Milk
  HECH46199010
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Hydrometer for lime milk without thermometer
  • Length:
    ~29 cm
  • Reference temperature:
    20 °C
  • Pk:
    1
  • Range:
    0 - 340: 5,0 g/ml
Hydrometer for lime milk without thermometer.