Anti-FRA1 (Ser265) Phosphate Rabbit Polyclonal Antibody
Catalog # CSTA3880S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:FRA1 (Ser265) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CSSDPLGsPTLLAL
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: FRA1 (Ser265) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat