Anti-Cyclin E1 Rabbit Monoclonal Antibody [clone: D7T3U]
Catalog # CSTA20808S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Cyclin E1
- Clonality:Monoclonal
- Clone:D7T3U
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CRETGSSKLKHFRGVADEDAH
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Cyclin E1
Clonality: Monoclonal
Clone: D7T3U
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat