Anti-Vimentin Rabbit Monoclonal Antibody [clone: D21H3]
Catalog # CSTA5741S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Vimentin
- Clonality:Monoclonal
- Clone:D21H3
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CTRTYSLGSALRPSTSRSLYA
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: D21H3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey