Anti-PKA C-alpha Rabbit Polyclonal Antibody
CSTA4782S
- Antibody type:Primary
- Antigen symbol:PKA C-alpha
- Clonality:Polyclonal
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CKGPGDTSNFDDYEE
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: PKA C-alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat