Anti-RagC Rabbit Monoclonal Antibody [clone: D8H5]
Catalog # CSTA9480S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:RagC
- Clonality:Monoclonal
- Clone:D8H5
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:SLQYGAEETPLAGSYGAADSC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RagC
Clonality: Monoclonal
Clone: D8H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey