Anti-CHOP Rabbit Monoclonal Antibody [clone: D46F1]
Catalog # CSTA5554S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CHOP
- Clonality:Monoclonal
- Clone:D46F1
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Seq:02
- Sequence:CRLTREVEATRRALIDRMVNLHQA
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CHOP
Clonality: Monoclonal
Clone: D46F1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse