Anti-Cyclin H Rabbit Polyclonal Antibody
CSTA2927S
- Antibody type:Primary
- Antigen symbol:Cyclin H
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:RKGYEDDDYVSKK(P/S)KC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Cyclin H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat