ContactUsLinkComponent
Height Endurance Vented Helmet
Height Endurance Vented Helmet
Catalog # PRTWPS63ORR
Supplier:  Portwest
Height Endurance Vented Helmet
Catalog # PRTWPS63ORR
Supplier:  Portwest
Specifications
  • Colour:
    Orange
  • Description:
    Height endurance vented helmet
  • Pk:
    1
Specifications
About this item
The premium Height Endurance Helmet is available in a vented version, for extra air flow. Light, comfortable, and compact, this peak-less helmet is designed to be used when working at heights.
  • Vented helmet shell allows air circulation, cooling the wearer's head
  • Six point textile suspension harness for enhanced wearer comfort and safety
  • Size adjustable headband via wheel ratchet system
  • Y-style, four-point chin strap
  • Hi-visibility colour for visibility in daylight
  • Up to seven years lifetime from manufacturing date (as long as undamaged)
Clever features include an ABS shell, 6-point textile suspension harness, wheel ratchet adjustment, size 52 to 63 cm. Technical sweatband and 4-point chin strap (Y-style) with a soft rubber chin protector are included.
Compatible with PW47 and PS47 clip-on ear muff. Also compatible with PS58 helmet visor holder.
Certifications: ANSI/ISEA Z89.1 TYPE I (Class C).