Order Entry
Northern Ireland
ContactUsLinkComponent
1,4-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Supelco®
1,4-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Supelco®
Catalog # 48049.
Supplier:  Merck
CAS Number:  
1,4-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Supelco®
Catalog # 48049.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    3855-82-1
  • Description:
    1,4-Dichlorobenzene-[D4] 2,000 µg/ml in methanol
  • Formula:
    C₆H₄Cl₂
  • Description:
    1,4-Dichlorobenzene-[D4]

Specifications

About this item