Order Entry
Northern Ireland
ContactUsLinkComponent
 
Neutral red 90% (dye content), Sigma-Aldrich®
 
undefined
Neutral red 90% (dye content), Sigma-Aldrich®
 

 

  • Environmentally Preferable:

 

 

  • Appearance (Colour):
    Brown to Black may have red or green sheen
  • Appearance (Form):
    Powder
  • Solubility (Colour), 10 mg/ml, H₂O:
    Dark Red
  • Infrared spectrum:
    Conforms to Structure
  • Dye Content (Calculated based on UV spectral assay):
    ≥90%
  • Recommended Retest Period:
    4 years

 

 

Frequently Bought Together