Neutral red, Sigma-Aldrich®
Glass bottle for solids, 25 g
Catalog # 861251-25G
Supplier: SIGMA ALDRICH MICROSCOPY
Neutral red, Sigma-Aldrich®
Catalog # 861251-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 553-24-2
Jump to:
Product Details & Documents
Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
- CAS Number:553-24-2
- Formula:C₁₅H₁₇ClN₄
- MDL Number:MFCD00012651
- Molecular Weight:288.78 g/mol
- Merck Index:12,06571
- Melting Pt:290 °C
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:209-035-8
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



