1-(Trimethylsilyl)imidazole, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:18156-74-6
:
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280
:140.26 g/mol
Boiling Pt:209 °C (1013 hPa)
Melting Pt:–42 °C
Density:0,95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:61 °C
EINECS:242-040-3
Frequently Bought Together