

Acetic acid glacial ≥99.7% ACS, Fluka™
320099-500ML
:
- Pack type:Glass bottle, narrow mouth
- Pk:500 mL
- Seq:2725
- Vapor Density:2.07 (vs air)
- Assay (GC):Min. 99.70 %
- Identity (IR):complying
- Non-volatile matter:Max. 0.001 %
- Mixable with H₂O:complying
- Titrable Base:Max. 0.0004 meq/g
- Vapor Pressure:11.4 mmHg ( 20 °C)
- Explosion Limit:16 %, 92 °F
- Iron (Fe):Max. 0.2 ppm
- Heavy metals (as Pb):Max. 0.5 ppm
- Chloride (Cl):Max. 1 ppm
- Sulfate (SO₄):Max. 1 ppm
- APHA:Max. 10
- Acetic aNHydride (GC):Max. 0.01 %
- K₂Cr₂O₇ reducing matter:complying
- KMnO₄ reducing substances:complying
:
:60.05 g/mol
:118 °C (1,013 hPa)
:16 °C
:1,05 g/cm³ (20 °C)
:40 °C
:Ambient
:200-580-7
:2789
:8,II
:01-2119475328-30
:GHS02
:GHS05
:64-19-7
:H₃CCOOH
:MFCD00036152