(±)-2-Methyltetrahydrofuran ≥99.5% stabilised, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:96-47-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00005367
Boiling Pt:78…80 °C (1.013 hPa)
Density:0.86 g/cm³ (20 °C)
:150-400 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Molecular Weight:86.13 g/mol
Melting Pt:<–20 °C
Flash Pt:–10 °C
Storage Temperature:Ambient
EINECS:202-507-4
UN:2536
ADR:3 II
:GHS07
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 99.50 %
- Assay BHT (GC):150 - 400 ppm
- Water (Karl Fischer):Max. 1.0 %
- IR spectrum:complying
- Peroxides (as H₂O₂):Max. 100 ppm
- Contains:150-400 ppm BHT as stabilizer
- Refractive Index:n20/D 1.406 (lit.)
Specification Test Results
Frequently Bought Together