4-Formylbenzoic acid
Catalog # MOLE17036513-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.13 g/mol
Boiling Pt:306 °C (1013 hPa)
Boiling Pt:306 °C (1013 hPa)
Melting Pt:148 °C
EINECS:210-607-4
CAS Number:619-66-9
Formula:C₈H₆O₃
MDL Number:MFCD00006951