Undec-10-enoic acid
Catalog # MOLEM27738770
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:184.28 g/mol
Merck Index:12,09983
Boiling Pt:300 °C (1013 hPa)
Melting Pt:24 °C
Density:0,91 g/cm³ (20 °C)
Flash Pt:146 °C
EINECS:203-965-8
:GHS07
CAS Number:112-38-9
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00004442