
Aluminium chloride, anhydrous, powder, Msynth® plus, Sigma-Aldrich®
8.45019.0100
: Merck
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Solubility:450 g/l (20 °C) (decomposition)
- Melting Point:180 - 181 °C (sublimed)
- Molar Mass:133.34 g/mol
- Bulk Density:1200 kg/m3
- Vapor Pressure:1 hPa (20 °C)
- Density:2.44 g/cm3 (20 °C)
- pH:2.4 (100 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive material
:
:133.34 g/mol
:13,00335
:180 °C (1013 hPa)
:180…181 °C
:liquid 1,31 g/cm³ (20 °C)
:Ambient
:231-208-1
:1726
:8,II
:M-Clarity™ quality level = MQ400
:GHS05
:7446-70-0
:AlCl₃
:MFCD00003422