Order Entry
Northern Ireland
ContactUsLinkComponent
3-Fluorophthalic acid for synthesis, Sigma-Aldrich®
Irritant
3-Fluorophthalic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41139.0001
Supplier:  Merck
CAS Number:  
Irritant
3-Fluorophthalic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41139.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Melting Point:
    158 - 161 °C
  • Molar Mass:
    184.12 g/mol

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :184.12 g/mol
EINECS:216-433-5
 :GHS07
CAS Number:1583-67-1
Formula:C₈H₅FO₄
MDL Number:MFCD00039524