4-Formylbenzoic acid, Sigma-Aldrich®
Catalog # 8.21075.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:248 - 250 °C
- Molar Mass:150.14 g/mol
- Bulk Density:396 kg/m3
- pH:3.5 (1 g/l, H2O, 20 °C)
- Synonyms:4-Formylbenzoic acid, 4-Carboxybenzaldehyde
- Hill Formula:C8H6O3
- Chemical Formula:4-(OHC)C6H4COOH
Specification Test Results
About this item
:
:150.13 g/mol
Boiling Pt:306 °C (1013 hPa)
Melting Pt:148 °C
:M-Clarity™ quality level = MQ200
EINECS:210-607-4
CAS Number:619-66-9
Formula:C₈H₆O₃
MDL Number:MFCD00006951