Order Entry
Northern Ireland
ContactUsLinkComponent
Ethoxytrimethylsilane for synthesis, Sigma-Aldrich®
Flammable
Irritant
Ethoxytrimethylsilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Ethoxytrimethylsilane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay (GC, area%):
    ≥ 97.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.756 - 0.759
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Frequently Bought Together