Bis(triphenylphosphine)copper(I) borohydride for synthesis, Sigma-Aldrich®
Catalog # 8.18504.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:160 - 165 °C decomposes
- Molar Mass:602.97 g/mol
Specification Test Results
About this item
Synonyms: Copper bis(triphenylphosphine)(tetrahydroborate), Copper bis(triphenylphosphine)(borhydride), Tris(triphenylphosphine)copper (I) tetrahydroborate
:
:602.97 g/mol
EINECS:240-951-0
UN:2813
:GHS07
Storage Temperature:Ambient
CAS Number:16903-61-0
Formula:C₃₆H₃₄BCuP₂
MDL Number:MFCD00013219
:M-Clarity™ quality level = MQ200