
(1-Butyl)triphenylphosphonium chloride for synthesis
Catalog # 8.14141.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:223 - 226 °C
- Molar Mass:354.85 g/mol
- Flash Point:270 °C
- Synonyms:Triphenylbutylphosphonium chloride
- Hill Formula:C22H24ClP
- Chemical Formula:(C6H5)3P+C4H9Cl-
Specification Test Results
About this item
:
:354.86 g/mol
Melting Pt:222…226 °C
Storage Temperature:Ambient
EINECS:236-443-3
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:>270 °C
UN:3464
:GHS07
CAS Number:13371-17-0
Formula:H₃C(CH₂)₃P(C₆H₅)₃Cl
MDL Number:MFCD00040546