Order Entry
Northern Ireland
ContactUsLinkComponent
3,3'-Thiodipropionic acid, Sigma-Aldrich®
3,3'-Thiodipropionic acid, Sigma-Aldrich®
Catalog # 8.08626.0250
Supplier:  Merck
CAS Number:  
3,3'-Thiodipropionic acid, Sigma-Aldrich®
Catalog # 8.08626.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
Merck Index:12,09468
Boiling Pt:347 °C (1013 hPa)
Melting Pt:128…131 °C
Density:1,496 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :178.21 g/mol
EINECS:203-841-3
CAS Number:111-17-1
Formula:C₆H₁₀O₄S
MDL Number:MFCD00002781