Order Entry
Northern Ireland
ContactUsLinkComponent
1,3-Dioxolane stabilised, Sigma-Aldrich®
Flammable
1,3-Dioxolane stabilised, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
1,3-Dioxolane stabilised, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -26 °C
  • Molar Mass:
    74.08 g/mol
  • Boiling Point:
    74 - 75 °C (1013 hPa)
  • Vapor Pressure:
    133 hPa (20 °C)
  • Flash Point:
    2 °C
  • Refractive Index:
    1.4 (20 °C, 589 nm)
  • Explosion Limit:
    2.1 - 20.5 %(V)
  • Density:
    1.065 g/cm3 (20 °C)
  • Ignition Point:
    274 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Frequently Bought Together